missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiophene-2-boronic acid, 96%, Acros Organics™

Brand:  Acros Organics 359600050

Code : 29

Additional Details : CAS Number : 162607-15-0 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10332144

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

4-Methylthiophene-2-boronic acid
95.0
≥95%
C5H7BO2S
4-Methyl-2-thienylboric acid
162607-15-0
100.0
96%
141.99

Specifications

4-Methylthiophene-2-boronic acid
5g
95.0
Authentic
Powder
96%
141.99
White to light beige
162607-15-0
100.0
Glass bottle
≥95%
C5H7BO2S
4-Methyl-2-thienylboric acid
Videos
SDS