missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Catalog Number p-3881719
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
958443-39-5
C10H8N2OS
204.247
MFCD12198119
KHHZRQVJLBGSNM-UHFFFAOYSA-N
4-methyl-2-pyridin-3-yl thiazole-5-carbaldehyde, 4-methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-thiazole-5-carbaldehyde, 4-methyl-2-3-pyridyl-1,3-thiazole-5-carbaldehyde
43811058
4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde
CC1=C(SC(=N1)C2=CN=CC=C2)C=O
This item is not returnable. View return policy
958443-39-5
C10H8N2OS
204.247
MFCD12198119
KHHZRQVJLBGSNM-UHFFFAOYSA-N
4-methyl-2-pyridin-3-yl thiazole-5-carbaldehyde, 4-methyl-2-pyrid-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-3-yl-thiazole-5-carbaldehyde, 4-methyl-2-3-pyridyl-1,3-thiazole-5-carbaldehyde
43811058
4-methyl-2-pyridin-3-yl-1,3-thiazole-5-carbaldehyde
CC1=C(SC(=N1)C2=CN=CC=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-pyrid-3-yl-1, 3-thiazole-5-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'