missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4510223
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
103249-79-2
C12H10N2O2
214.224
ZPXPNKCBUXSJFK-UHFFFAOYSA-N
4-methyl-2-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 4-methyl-2-phenyl, acmc-1c8q0, 2-phenyl-4-methylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid,4-methyl-2-phenyl, 4-methyl-2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 4-methyl-2-phenyl
239975
4-methyl-2-phenylpyrimidine-5-carboxylic acid
CC1=NC(=NC=C1C(=O)O)C2=CC=CC=C2
This item is not returnable. View return policy
103249-79-2
C12H10N2O2
214.224
ZPXPNKCBUXSJFK-UHFFFAOYSA-N
4-methyl-2-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 4-methyl-2-phenyl, acmc-1c8q0, 2-phenyl-4-methylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid,4-methyl-2-phenyl, 4-methyl-2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 4-methyl-2-phenyl
239975
4-methyl-2-phenylpyrimidine-5-carboxylic acid
CC1=NC(=NC=C1C(=O)O)C2=CC=CC=C2
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'