missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride, Technical, Thermo Scientific™

Catalog Number 10641275
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
690632-88-3
C10H8ClNO2S2
273.749
NGDQQLAVJWUYSF-UHFFFAOYSA-N
4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride
2795490
4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl
This item is not returnable. View return policy
690632-88-3
C10H8ClNO2S2
273.749
NGDQQLAVJWUYSF-UHFFFAOYSA-N
4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride
2795490
4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 273.76
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'