missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methanol, 97%, Thermo Scientific™

Catalog Number 10710354
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
61291-91-6
C11H11NOS
205.275
MFCD01928782
JSIPFWSPCWZJIU-UHFFFAOYSA-N
4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole
736541
(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol
CC1=C(SC(=N1)C2=CC=CC=C2)CO
This item is not returnable. View return policy
61291-91-6
C11H11NOS
205.275
MFCD01928782
JSIPFWSPCWZJIU-UHFFFAOYSA-N
4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole
736541
(4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol
CC1=C(SC(=N1)C2=CC=CC=C2)CO
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.28
Percent Purity 97%
Chemical Name or Material (4-Methyl-2-phenyl-1, 3-thiazol-5-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'