missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-2-phenyl-1,3-oxazole-5-carbaldehyde, 95%, Maybridge™
£71.33 - £194.00
Chemical Identifiers
CAS | 953408-85-0 |
---|---|
Molecular Formula | C11H9NO2 |
Molecular Weight (g/mol) | 187.198 |
MDL Number | MFCD11109318 |
InChI Key | NIYJTYPVRBYCEZ-UHFFFAOYSA-N |
Synonym | 5-formyl-4-methyl-2-phenyl-1,3-oxazole, 4-methyl-2-phenyloxazole-5-carbaldehyde, 4-methyl-2-phenyl-1,3-oxazole-5-carboxaldehyde |
PubChem CID | 26343583 |
IUPAC Name | 4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde |
SMILES | CC1=C(OC(=N1)C2=CC=CC=C2)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10021124
|
Maybridge
CC18104CB |
250mg | Amber Glass Bottle |
£71.33
250mg |
Estimated Shipment 04-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10763995
|
Maybridge
CC18104DA |
1g | Amber Glass Bottle |
£194.00
1g |
Estimated Shipment 04-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Identificadores de productos químicos
953408-85-0 | |
187.198 | |
NIYJTYPVRBYCEZ-UHFFFAOYSA-N | |
26343583 | |
CC1=C(OC(=N1)C2=CC=CC=C2)C=O |
C11H9NO2 | |
MFCD11109318 | |
5-formyl-4-methyl-2-phenyl-1,3-oxazole, 4-methyl-2-phenyloxazole-5-carbaldehyde, 4-methyl-2-phenyl-1,3-oxazole-5-carboxaldehyde | |
4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde |
Especificaciones
4-Methyl-2-phenyl-1, 3-oxazole-5-carbaldehyde | |
250mg | |
94.5 | |
95% | |
MFCD11109318 | |
NIYJTYPVRBYCEZ-UHFFFAOYSA-N | |
4-methyl-2-phenyl-1,3-oxazole-5-carbaldehyde | |
26343583 | |
95% |
Amber Glass Bottle | |
953408-85-0 | |
100.0 | |
C11H9NO2 | |
5-formyl-4-methyl-2-phenyl-1,3-oxazole, 4-methyl-2-phenyloxazole-5-carbaldehyde, 4-methyl-2-phenyl-1,3-oxazole-5-carboxaldehyde | |
CC1=C(OC(=N1)C2=CC=CC=C2)C=O | |
187.198 | |
187.2 |