(4-Methyl-2-phenyl-1,3-oxazol-5-ylmethyl)amine, 97%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: CC18113DA

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 165735-97-7 Weight: 0.05100kg


Packaging Amber Glass Bottle
Quantity 1g

Product Code. 10162974

Quantity Price
1 £ 199.0 / 1g
Estimated Shipment date
from Supplier 31-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (4-Methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine
CAS 165735-97-7
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12N2O
Formula Weight 188.23
MDL Number MFCD11109320

Specification

Formula Weight 188.23
Quantity 1g
Assay 97%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
CAS Min % 96.5
CAS 165735-97-7
MDL Number MFCD11109320
Molecular Formula C11H12N2O
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material (4-Methyl-2-phenyl-1, 3-oxazol-5-ylmethyl)amine
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.