missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, 99.5%, for analysis, ACROS Organics™

Brand:  Acros Organics 255660250

Code : 29

Additional Details : CAS Number : 108-10-1 Weight : 20.08000kg Transport : UN number : 1245 Chem class : 3 Pack group : II

Quantity 25L
Packaging Metal drum

 View more versions of this product

Product Code. 10503434

  • £321.70 / 25 L


Estimated Shipment
16-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Methyl-2-pentanone
99.4
For analysis
99.5%
(CH3)2CHCH2COCH3
MFCD00008938
108-10-1
100.0
99.5%
C6H12O
100.16
Isobutyl methyl ketone, Isopropylacetone, Methyl isobutyl ketone, MIBK

Specifications

0.0004 meq/g max.
117.4°C
108-10-1
100.0
0.1ppm max.
0.02ppm max.
0.02ppm max.
Authentic
For analysis
Metal drum
99.5%
0.005% max.
C6H12O
100.16
0.1ppm max.
0.8000g/mL
Clear
14°C
4-Methyl-2-pentanone
25L
99.4
0.05ppm max.
0.5ppm max.
0.02ppm max.
0.1ppm max.
0.1ppm max.
1.3950 to 1.3970
Liquid
99.5%
0.8
(CH3)2CHCH2COCH3
MFCD00008938
0.2% max.
Isobutyl methyl ketone, Isopropylacetone, Methyl isobutyl ketone, MIBK
− 84.0°C
Videos
SDS