missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(4-(trifluoromethyl)phenyl)-1,3-thiazole-5-sulfonyl chloride, 90%, Thermo Scientific™

Catalog Number p-3075647
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
568577-83-3
C11H7ClF3NO2S2
341.747
MFCD04974054
FIPBWMJINRBLKG-UHFFFAOYSA-N
4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-sulfonyl chloride, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-sulfonyl chloride
2794632
4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)S(=O)(=O)Cl
This item is not returnable. View return policy
568577-83-3
C11H7ClF3NO2S2
341.747
MFCD04974054
FIPBWMJINRBLKG-UHFFFAOYSA-N
4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-sulfonyl chloride, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-sulfonyl chloride
2794632
4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-sulfonyl chloride
CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)S(=O)(=O)Cl
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 341.76
Percent Purity 90%
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'