missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthaleneboronic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-4259022
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
103986-53-4
C11H11BO2
186.02
MFCD01632204
JHVQEUGNYSVSDH-UHFFFAOYSA-N
4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl
2773511
(4-methylnaphthalen-1-yl)boronic acid
CC1=CC=C(B(O)O)C2=CC=CC=C12
This item is not returnable. View return policy
103986-53-4
C11H11BO2
186.02
MFCD01632204
JHVQEUGNYSVSDH-UHFFFAOYSA-N
4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl
2773511
(4-methylnaphthalen-1-yl)boronic acid
CC1=CC=C(B(O)O)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 224°C to 228°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 186.02
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Methyl-1-naphthaleneboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-1-naphthaleneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'