missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-1-indanone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 98°C to 100°C |
Color | White to Yellow |
Boiling Point | 144°C (13.0 Torr) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Quantity | 5 g |
Formula Weight | 146.19 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'