missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-1,3-oxazole-5-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 127.1 |
Percent Purity | 95% |
Chemical Name or Material | 4-Methyl-1, 3-oxazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'