missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypyridine, 99%, ACROS Organics™

Brand:  Acros Organics 379940250

Code : 29

Additional Details : CAS Number : 620-08-6 Weight : 0.07688kg


Quantity 25mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10341163

  • £174.80 / 25mL


Estimated Shipment
11-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Methoxypyridine
98.5
99%
C6H7NO
MFCD00674049
620-08-6
100.0
99%
109.13

Specifications

4-Methoxypyridine
98.5
Glass bottle
1.5180 to 1.5220
1.075
99%
1.0750g/mL
109.13
191.0°C
21, V,02, 98
25mL
620-08-6
100.0
Authentic
Darkens on storage
Liquid
99%
C6H7NO
MFCD00674049
Clear colorless to slightly yellow
4.0°C
74°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Methoxypyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS