missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an organometallic compounds are widely used as intermediates in organic synthesis.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong ba
It is an organometallic compounds are widely used as intermediates in organic synthesis.
Solubility
Reacts with water.
Notes
Handle under dry protective gas. Air sensitive. Store away from strong ba
Specifications
CAS | 254454-47-2 |
Density | 1.006 |
Molecular Formula | C7H7IOZn |
MDL Number | MFCD01311444 |
Quantity | 50 mL |
UN Number | UN3399 |
Beilstein | 8478508 |
Sensitivity | Air and light sensitive |
Synonym | zinc 2+ ion anisole iodide |
Solubility Information | Reacts with water. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'