missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals

Catalog Number 15403007
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
13139-86-1
C7H7BrMgO
211.34
MFCD00013515
JKXAZEXIOPFJSC-UHFFFAOYSA-M
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
2734898
COC1=CC=C([Mg]Br)C=C1
This item is not returnable. View return policy
13139-86-1
C7H7BrMgO
211.34
MFCD00013515
JKXAZEXIOPFJSC-UHFFFAOYSA-M
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
2734898
COC1=CC=C([Mg]Br)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Density 0.955 g/mL
Flash Point -20°C (-4°F)
Quantity 500 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name magnesium;methoxybenzene;bromide
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 0.5M in THF
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'