missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog Number p-3758326
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7693-41-6
C8H7ClO3
186.591
MFCD00013258
CCFSGQKTSBIIHG-UHFFFAOYSA-N
4-methoxyphenyl chloroformate, carbonochloridic acid, 4-methoxyphenyl ester, 4-methoxyphenylchloroformate, chloroformic acid 4-methoxyphenyl ester, acmc-1bfyr, p-methoxyphenyl chloroformate, 4-methoxy-phenyl chloroformate, 4-methoxy phenyl chloroformate, 4-methoxyphenyl carbonochloridate
82128
(4-methoxyphenyl) carbonochloridate
COC1=CC=C(C=C1)OC(=O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7693-41-6
C8H7ClO3
186.591
MFCD00013258
CCFSGQKTSBIIHG-UHFFFAOYSA-N
4-methoxyphenyl chloroformate, carbonochloridic acid, 4-methoxyphenyl ester, 4-methoxyphenylchloroformate, chloroformic acid 4-methoxyphenyl ester, acmc-1bfyr, p-methoxyphenyl chloroformate, 4-methoxy-phenyl chloroformate, 4-methoxy phenyl chloroformate, 4-methoxyphenyl carbonochloridate
82128
(4-methoxyphenyl) carbonochloridate
COC1=CC=C(C=C1)OC(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.25g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula ClCOC6H4OCH3
Refractive Index 1.5220 to 1.5260 (20°C, 589nm)
Quantity 5 g
UN Number 3277
Formula Weight 186.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl chloroformate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'