missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride, 97%, Thermo Scientific™

Catalog Number p-2695137
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
25519-82-8
C13H18ClNO2
255.742
MFCD00114817
BBDTWYQCXXFKDH-UHFFFAOYSA-N
4-4-methoxybenzoyl piperidine hydrochloride, 4-methoxyphenyl 4-piperidyl methanone hydrochloride, 4-methoxyphenyl piperidin-4-yl methanone hydrochloride, methanone, 4-methoxyphenyl-4-piperidinyl-, hydrochloride, 4-4-methoxybenzoyl piperidine hcl, 4-4-methoxybenzoyl-piperidine hydrochloride, 4-methoxy-phenyl-piperidin-4-yl-methanone hydrochloride, ksc801q4b, 4-methoxyphenyl-4-piperidyl methanonehcl, 4-methoxyphenyl 4-piperidyl ketone, chloride
2740587
(4-methoxyphenyl)-piperidin-4-ylmethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)C2CCNCC2.Cl
This item is not returnable. View return policy
25519-82-8
C13H18ClNO2
255.742
MFCD00114817
BBDTWYQCXXFKDH-UHFFFAOYSA-N
4-4-methoxybenzoyl piperidine hydrochloride, 4-methoxyphenyl 4-piperidyl methanone hydrochloride, 4-methoxyphenyl piperidin-4-yl methanone hydrochloride, methanone, 4-methoxyphenyl-4-piperidinyl-, hydrochloride, 4-4-methoxybenzoyl piperidine hcl, 4-4-methoxybenzoyl-piperidine hydrochloride, 4-methoxy-phenyl-piperidin-4-yl-methanone hydrochloride, ksc801q4b, 4-methoxyphenyl-4-piperidyl methanonehcl, 4-methoxyphenyl 4-piperidyl ketone, chloride
2740587
(4-methoxyphenyl)-piperidin-4-ylmethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)C2CCNCC2.Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 255.74
Percent Purity 97%
Chemical Name or Material (4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'