missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenethyl alcohol, 96%, Thermo Scientific Chemicals

Catalog Number p-157716
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
702-23-8
C9H12O2
152.19
MFCD00002900
AUWDOZOUJWEPBA-UHFFFAOYSA-N
2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
69705
2-(4-methoxyphenyl)ethanol
COC1=CC=C(C=C1)CCO
This item is not returnable. View return policy
702-23-8
C9H12O2
152.19
MFCD00002900
AUWDOZOUJWEPBA-UHFFFAOYSA-N
2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
69705
2-(4-methoxyphenyl)ethanol
COC1=CC=C(C=C1)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 28°C to 30°C
Color Brown
Boiling Point 334°C to 336°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CH2OH
Refractive Index 1.5360 to 1.538
Quantity 10 g
Beilstein 06, 906
Formula Weight 152.19
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 4-Methoxyphenethyl alcohol, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'