missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxynaphthalene-1-carboxylic acid, 98%

Brand:  Alfa Aesar™ H64432.06

Code : NEW

Additional Details : CAS Number : 13041-62-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451639

  • £104.90 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Description

Description

Applications
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

4-Methoxynaphthalene-1-carboxylic acid
202.21
2099800
13041-62-8
MFCD02575372
4-Methoxy-1-naphthoic acid

Specifications

4-Methoxynaphthalene-1-carboxylic acid
243° to 247°C
13041-62-8
98%
MFCD02575372
4-Methoxy-1-naphthoic acid
Off-white
5g
Crystalline Solid
202.21
2099800
Videos
SDS