missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxymetanilyl fluoride, 97%, Thermo Scientific™

Catalog Number p-157704
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
498-74-8
C7H8FNO3S
205.2
MFCD00007417
SESLQGPHIQXYGF-UHFFFAOYSA-N
4-methoxymetanilyl fluoride, 3-amino-4-methoxyphenylsulphonyl fluoride, 3-amino-4-methoxy benzene sulphonylfluoride, 3-amino-4-methoxyphenyl fluorosulfone, 3-amino-4-methoxy-benzenesulfonyl fluoride, 3-amino-4-methoxybenzenesulphonyl fluoride, 3-azanyl-4-methoxy-benzenesulfonyl fluoride, benzenesulfonylfluoride, 3-amino-4-methoxy, 3-amino-4-methoxybenzene-1-sulfonyl fluoride, benzenesulfonyl fluoride, 3-amino-4-methoxy
96952
3-amino-4-methoxybenzenesulfonyl fluoride
COC1=C(C=C(C=C1)S(=O)(=O)F)N
This item is not returnable. View return policy
498-74-8
C7H8FNO3S
205.2
MFCD00007417
SESLQGPHIQXYGF-UHFFFAOYSA-N
4-methoxymetanilyl fluoride, 3-amino-4-methoxyphenylsulphonyl fluoride, 3-amino-4-methoxy benzene sulphonylfluoride, 3-amino-4-methoxyphenyl fluorosulfone, 3-amino-4-methoxy-benzenesulfonyl fluoride, 3-amino-4-methoxybenzenesulphonyl fluoride, 3-azanyl-4-methoxy-benzenesulfonyl fluoride, benzenesulfonylfluoride, 3-amino-4-methoxy, 3-amino-4-methoxybenzene-1-sulfonyl fluoride, benzenesulfonyl fluoride, 3-amino-4-methoxy
96952
3-amino-4-methoxybenzenesulfonyl fluoride
COC1=C(C=C(C=C1)S(=O)(=O)F)N

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)SO2F
Quantity 10 g
Beilstein 14, II, 487
Formula Weight 205.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxymetanilyl fluoride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxymetanilyl fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'