missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Methoxychalcone, 97%, Thermo Scientific Chemicals
16.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.
Solubility
Soluble in dichloromethane and methanol. Insoluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.
Solubility
Soluble in dichloromethane and methanol. Insoluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
Specifications
Melting Point | 101°C to 103°C |
Quantity | 5 g |
Beilstein | 644640 |
Solubility Information | Soluble in dichloromethane and methanol. Insoluble in water. |
Formula Weight | 238.29 |
Percent Purity | 97% |
Chemical Name or Material | 4'-Methoxychalcone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'