missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxybenzyl)triphenylphosphonium chloride, 98%, Thermo Scientific™

Catalog Number p-3593195
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
3462-97-3
C26H24ClOP
418.90
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
2773651
[Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
This item is not returnable. View return policy
3462-97-3
C26H24ClOP
418.90
MFCD00031572
YQXBNCFNXOFWLR-UHFFFAOYSA-M
4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
2773651
[Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 244.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2P(C6H5)3Cl
Quantity 5 g
IUPAC Name [(4-methoxyphenyl)methyl]triphenylphosphanium chloride
Formula Weight 418.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (4-Methoxybenzyl)triphenylphosphonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • (4-Methoxybenzyl)triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'