missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-156951
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
611-94-9
C14H12O2
212.25
MFCD00008403
SWFHGTMLYIBPPA-UHFFFAOYSA-N
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
69146
(4-methoxyphenyl)-phenylmethanone
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
This item is not returnable. View return policy
611-94-9
C14H12O2
212.25
MFCD00008403
SWFHGTMLYIBPPA-UHFFFAOYSA-N
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
69146
(4-methoxyphenyl)-phenylmethanone
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 63°C
Color Orange-Yellow to White
Boiling Point 354°C to 356°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COC6H5
Quantity 25 g
Beilstein 08, 159
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol and ether
Formula Weight 212.25
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxybenzophenone, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'