missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl Chloride, 99%, Thermo Scientific Chemicals

Catalog Number 10524274
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
98-68-0
C7H7ClO3S
206.64
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
98-68-0
C7H7ClO3S
206.64
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(Cl)(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 40.0°C to 42.0°C
Color White to Beige
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4SO2Cl
Quantity 500 g
Beilstein 11, 243
Fieser 09,297
Solubility Information Solubility in water: decomposes
Formula Weight 206.65
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxybenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'