missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021453
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
98-68-0
C7H7ClO3S
206.64
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
98-68-0
C7H7ClO3S
206.64
MFCD00007446
DTJVECUKADWGMO-UHFFFAOYSA-N
4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
7401
4-methoxybenzenesulfonyl chloride
COC1=CC=C(C=C1)S(Cl)(=O)=O

4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.

Solubility
Soluble in Toluene. Decomposes in water.

Notes
Moisture Sensitive. Store at 4°C. Store in a cool and dark place. Protect from moisture. Store away from strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 37°C to 42°C
Boiling Point 119°C to 122°C (0.4 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 609005
Sensitivity Moisture sensitive
Solubility Information Soluble in Toluene. Decomposes in water.
Formula Weight 206.65
Percent Purity ≥98%
Chemical Name or Material 4-Methoxybenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'