missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzamidoxime, 97%, Thermo Scientific Chemicals

Catalog Number p-7034456
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5373-87-5
C8H10N2O2
166.18
MFCD05664434
WVALRFKCJCIVBR-UHFFFAOYSA-N
4-methoxybenzamidoxime, z-n'-hydroxy-4-methoxybenzimidamide, p-methoxybenzamidoxime, n-hydroxy-4-methoxybenzenecarboximidamide, z-n'-hydroxy-4-methoxybenzene-1-carboximidamide, n-hydroxy-4-methoxy-benzamidine, n'-hydroxy-4-methoxybenzimidamide, hydroxyimino 4-methoxyphenyl methylamine, benzenecarboximidamide, n-hydroxy-4-methoxy, p-methoxybenzamideoxime
5360053
N'-hydroxy-4-methoxybenzenecarboximidamide
COC1=CC=C(C=C1)C(=NO)N
This item is not returnable. View return policy
5373-87-5
C8H10N2O2
166.18
MFCD05664434
WVALRFKCJCIVBR-UHFFFAOYSA-N
4-methoxybenzamidoxime, z-n'-hydroxy-4-methoxybenzimidamide, p-methoxybenzamidoxime, n-hydroxy-4-methoxybenzenecarboximidamide, z-n'-hydroxy-4-methoxybenzene-1-carboximidamide, n-hydroxy-4-methoxy-benzamidine, n'-hydroxy-4-methoxybenzimidamide, hydroxyimino 4-methoxyphenyl methylamine, benzenecarboximidamide, n-hydroxy-4-methoxy, p-methoxybenzamideoxime
5360053
N'-hydroxy-4-methoxybenzenecarboximidamide
COC1=CC=C(C=C1)C(=NO)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 4-Methoxybenzamidoxime

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'