missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzamide, 97%, Thermo Scientific Chemicals

Catalog Number 10666322
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3424-93-9
C8H9NO2
151.16
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N
This item is not returnable. View return policy
3424-93-9
C8H9NO2
151.16
MFCD00007995
GUCPYIYFQVTFSI-UHFFFAOYSA-N
p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide
76959
4-methoxybenzamide
COC1=CC=C(C=C1)C(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 164.0°C to 167.0°C
Color White
Boiling Point 295.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4CONH2
Quantity 25 g
Beilstein 10, II, 100
Solubility Information Solubility in water: soluble
Formula Weight 151.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'