missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-o-phenylenediamine dihydrochloride, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
Specifications
Melting Point | ∼219°C (decomposition) |
Quantity | 1 g |
Beilstein | 3699636 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 211.09 |
Percent Purity | ≥97% |
Chemical Name or Material | 4-Methoxy-o-phenylenediamine dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'