missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-N-(2,5-dimethoxyphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59113.03

Code : NEW

Additional Details : CAS Number : 313366-86-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415328

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Methoxy-N-(2,5-dimethoxyphenyl)benzamide
313366-86-8
287.32
MFCD00686771

Specifications

4-Methoxy-N-(2,5-dimethoxyphenyl)benzamide
313366-86-8
97%
MFCD00686771
1g
Solid
287.32
Videos
SDS