missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-N-(2,3-dimethylphenyl)benzamide, 97%

Brand:  Alfa Aesar™ H59446.03

Code : NEW

Additional Details : CAS Number : 127291-96-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15485768

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Methoxy-N-(2,3-dimethylphenyl)benzamide
255.32
3551251
127291-96-7
MFCD00027542

Specifications

4-Methoxy-N-(2,3-dimethylphenyl)benzamide
127291-96-7
97%
MFCD00027542
1g
Solid
255.32
3551251
Videos
SDS