missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 97%, Thermo Scientific Chemicals

Catalog Number p-157688
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
82-57-5
MFCD00005008
NZVQLVGOZRELTG-UHFFFAOYSA-N
visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
6716
CHEBI:10002
4-methoxy-7-methylfuro[3,2-g]chromen-5-one
CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3
This item is not returnable. View return policy
82-57-5
MFCD00005008
NZVQLVGOZRELTG-UHFFFAOYSA-N
visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
6716
CHEBI:10002
4-methoxy-7-methylfuro[3,2-g]chromen-5-one
CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 142°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C13H10O4
Quantity 5 g
Merck Index 15, 10209
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in chloroform,sparingly soluble in alcohol
Molecular Weight (g/mol) 230.22
Formula Weight 230.22
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-7-methyl-5H-furo¢3,2-g!¢1!benzopyran-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'