missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-6-methyl-2H-pyran-2-one, 97%

Brand:  Alfa Aesar™ H61395.MD

Code : NEW

Additional Details : CAS Number : 672-89-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15427928

  • £10.00 / 250mg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Description

Description

Applications
4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

Solubility
Poorly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4-Methoxy-6-methyl-2H-pyran-2-one
672-89-9
140.14
MFCD00006640

Specifications

4-Methoxy-6-methyl-2H-pyran-2-one
250mg
Solid
140.14
83° to 86°C
672-89-9
97%
MFCD00006640
Videos
SDS