missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-pyridineboronic acid hydrate, 97%, Thermo Scientific Chemicals

Catalog Number 10609804
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1266615-79-5, 355004-67-0
C6H8BNO3
152.94
MFCD07186213
YUTPAZKVEOJQCY-UHFFFAOYSA-N
4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid
2762556
(4-methoxypyridin-3-yl)boronic acid
COC1=CC=NC=C1B(O)O
This item is not returnable. View return policy
1266615-79-5, 355004-67-0
C6H8BNO3
152.94
MFCD07186213
YUTPAZKVEOJQCY-UHFFFAOYSA-N
4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid
2762556
(4-methoxypyridin-3-yl)boronic acid
COC1=CC=NC=C1B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 152.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methoxy-3-pyridineboronic acid hydrate
compliance-icons
Product Identifier
  • 4-Methoxy-3-pyridineboronic acid hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'