missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-nitrobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 3-nitro-4-methoxybenzonitrile, 4-methoxy-3-nitro-benzonitrile, benzonitrile, 4-methoxy-3-nitro, 4-methoxy-3-nitrobenzenecarbonitrile, acmc-1cmtm, 5-cyano-2-methoxynitrobenzene, benzonitrile,4-methoxy-3-nitro |
Formula Weight | 178.15 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxy-3-nitrobenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'