missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobenzoic acid, 98%, Thermo Scientific™

Catalog Number 10702811
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
89-41-8
C8H7NO5
197.15
MFCD00007256
ANXBDAFDZSXOPQ-UHFFFAOYSA-N
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
66640
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
This item is not returnable. View return policy
89-41-8
C8H7NO5
197.15
MFCD00007256
ANXBDAFDZSXOPQ-UHFFFAOYSA-N
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid
66640
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-]
CAS Min % 97.5
CAS Max % 100.0
Melting Point 189°C to 194°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)CO2H
Quantity 100 g
Beilstein 10, 181
Formula Weight 197.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-nitrobenzoic acid, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'