missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxy-3'-nitroacetophenone, 98+%, Thermo Scientific™

Catalog Number p-7058286
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
6277-38-9
C9H9NO4
195.17
MFCD00017023
VXLKYQQBEPCMJE-UHFFFAOYSA-N
1-4-methoxy-3-nitrophenyl ethanone, 1-4-methoxy-3-nitrophenyl ethan-1-one, 4-methoxy-3-nitroacetophenone, 3'-nitro-4'-methoxyacetophenone, 4'-methoxy-3'-nitroacetophenone, 4-acetyl-2-nitroanisole, 4-methoxy-3-nitro-acetophenone, 1-acetyl-4-methoxy-3-nitrobenzene, 1-4-methoxy-3-nitrophenyl-ethanone
80470
1-(4-methoxy-3-nitrophenyl)ethanone
COC1=CC=C(C=C1[N+]([O-])=O)C(C)=O
This item is not returnable. View return policy
6277-38-9
C9H9NO4
195.17
MFCD00017023
VXLKYQQBEPCMJE-UHFFFAOYSA-N
1-4-methoxy-3-nitrophenyl ethanone, 1-4-methoxy-3-nitrophenyl ethan-1-one, 4-methoxy-3-nitroacetophenone, 3'-nitro-4'-methoxyacetophenone, 4'-methoxy-3'-nitroacetophenone, 4-acetyl-2-nitroanisole, 4-methoxy-3-nitro-acetophenone, 1-acetyl-4-methoxy-3-nitrobenzene, 1-4-methoxy-3-nitrophenyl-ethanone
80470
1-(4-methoxy-3-nitrophenyl)ethanone
COC1=CC=C(C=C1[N+]([O-])=O)C(C)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 97°C to 99°C
Quantity 10 g
Beilstein 646069
Formula Weight 195.18
Percent Purity ≥98%
Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'