missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxy-3-methylbenzaldehyde, 98%

Brand:  Alfa Aesar™ H64207.03

Code : NEW

Additional Details : CAS Number : 32723-67-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15431309

  • £28.41 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Methoxy-3-methylbenzaldehyde
150.18
2084045
32723-67-4
MFCD00003375
3-Methyl-p-anisaldehyde

Specifications

4-Methoxy-3-methylbenzaldehyde
80° to 85°C (1mmHg)
1g
1.5690
98%
MFCD00003375
3-Methyl-p-anisaldehyde
1.025g/mL
Pale yellow
32723-67-4
Liquid
150.18
2084045
Videos
SDS