Learn More
4-Methoxy-2-methylbenzaldehyde, 95%, ACROS Organics™
Brand: Acros Organics 427020010
Code : 29
Additional Details : CAS Number : 52289-54-0 Weight : 0.05100kg
Quantity | 1g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
4-Methoxy-2-methylbenzaldehyde | |
96.0 | |
≥96.0% | |
C9H10O2 | |
150.18 |
52289-54-0 | |
100.0 | |
97% | |
CH30C6H3(CH3)CHO | |
MFCD02261771 |
Specifications
4-Methoxy-2-methylbenzaldehyde | |
Clear colorless to yellow | |
52289-54-0 | |
100.0 | |
Authentic | |
Liquid | |
97% | |
CH30C6H3(CH3)CHO | |
MFCD02261771 |
1.1070g/mL | |
1g | |
96.0 | |
Consistent with structure | |
Glass bottle | |
≥96.0% | |
C9H10O2 | |
150.18 |