missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog Number p-7055658
20.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52289-54-0
C9H10O2
150.177
MFCD02261771
WICYVKGMEJSDAO-UHFFFAOYSA-N
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
283285
4-methoxy-2-methylbenzaldehyde
CC1=C(C=CC(=C1)OC)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
52289-54-0
C9H10O2
150.177
MFCD02261771
WICYVKGMEJSDAO-UHFFFAOYSA-N
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
283285
4-methoxy-2-methylbenzaldehyde
CC1=C(C=CC(=C1)OC)C=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.107 g/mL
Boiling Point 262°C to 267°C (literature)
Flash Point >110°C (230°F)
Refractive Index 1.571
Quantity 1 g
Beilstein 1933786
Formula Weight 150.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-Methoxy-2-methylbenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'