4-Methoxy-1-naphthaldehyde 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 146940100

10GR 4-Methoxy-1-naphthaldehyde, 99%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 15971-29-6 Weight: 0.06000kg


Packaging Glass bottle
Quantity 10g

Product Code. 10447160

Quantity Price
1 £ 33.8 / 10g
Estimated Shipment date
from Supplier 08-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Name Note 99%
CAS 15971-29-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H10O2
Linear Formula CH3OC10H6CHO
Formula Weight 186.21
MDL Number MFCD00004006

Specification

Assay ≥98.5%
Physical Form Low Melting Solid
Formula Weight 186.21
Quantity 10g
Color Slightly beige to pink-brown
Beilstein 08, 147
CAS Min % 98.5
EINECS Number 240-109-2
Linear Formula CH3OC10H6CHO
Infrared Spectrum Authentic
CAS 15971-29-6
Assay Percent Range 99%
MDL Number MFCD00004006
Packaging Glass bottle
Name Note 99%
Molecular Formula C12H10O2
CAS Max % 100.0
Chemical Name or Material 4-Methoxy-1-naphthaldehyde
Melting Point 32° to 35°C
Documentation

Technical documents