missing translation for 'onlineSavingsMsg'
Learn More

4-Meth-5-(4,4,5,5-tetrameth-1,3,2-dioxaborol-2-yl)-2-(4-(triflmeth)phen)-1,3-thiaz, 97%, Maybridge™

£32.15 - £102.00

Chemical Identifiers

CAS 690631-96-0
Molecular Formula C17H19BF3NO2S
Molecular Weight (g/mol) 369.209
MDL Number MFCD05664416
InChI Key ZUROXICAUJQTIB-UHFFFAOYSA-N
Synonym 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
PubChem CID 2794643
IUPAC Name 4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=C(C=C3)C(F)(F)F)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10014292
SDS View Documents
Maybridge
MO00919CB
250mg Amber Glass Bottle
£32.15
250mg
View alternative products
Save Up to 
»
Estimated Shipment
01-03-2021
Please to purchase this item. Need a web account? Register with us today!
10370453
SDS View Documents
Maybridge
MO00919DA
1g Amber Glass Bottle
£102.00
1g
View alternative products
Save Up to 
»
Estimated Shipment
01-03-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

690631-96-0
369.209
ZUROXICAUJQTIB-UHFFFAOYSA-N
2794643
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=C(C=C3)C(F)(F)F)C
C17H19BF3NO2S
MFCD05664416
4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole

Specifications

4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
250mg
97.0
C17H19BF3NO2S
4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl thiazole, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-boronic acid pinacol ester, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole, 4-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl ph-1,3-thiazole, 4-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-4-trifluoromethyl phenyl-1,3-thiazole
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(S2)C3=CC=C(C=C3)C(F)(F)F)C
369.209
369.21
Amber Glass Bottle
690631-96-0
97%
MFCD05664416
ZUROXICAUJQTIB-UHFFFAOYSA-N
4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole
2794643
97%
SDS