missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptopyridine, 96%, ACROS Organics™

Brand:  Acros Organics 164710500

Code : 29

Additional Details : CAS Number : 4556-23-4 Weight : 0.10000kg


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 10609843

  • £246.70 / 50g


Estimated Shipment
18-07-2018
Add to basket


Specifications

Chemical Identifiers

4-Mercaptopyridine
95.0
≥95%
C5H5NS
111.16
4556-23-4
100.0
96%
(C5H4N)SH
MFCD00006422

Specifications

4-Mercaptopyridine
95.0
Authentic
Crystalline Powder
96%
(C5H4N)SH
MFCD00006422
4-Pyridinethiol
50g
4556-23-4
100.0
Glass bottle
≥95%
C5H5NS
111.16
21, II, 35
Yellow to yellow-green or brown
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Mercaptopyridine

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS