missing translation for 'onlineSavingsMsg'
Learn More

4-(Maleimidomethyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7034593
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1315276-43-7
C17H20BNO4
313.16
MFCD18827482
DROVAVNVMSFFHB-UHFFFAOYSA-N
4-maleimidomethyl benzeneboronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrole-2,5-dione, amtb376, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl maleimide, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-1h-pyrrole-2,5-dione, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-pyrrole-2,5-dione
71306538
1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrole-2,5-dione
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3C(=O)C=CC3=O
This item is not returnable. View return policy
1315276-43-7
C17H20BNO4
313.16
MFCD18827482
DROVAVNVMSFFHB-UHFFFAOYSA-N
4-maleimidomethyl benzeneboronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl pyrrole-2,5-dione, amtb376, n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl maleimide, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-1h-pyrrole-2,5-dione, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-pyrrole-2,5-dione
71306538
1-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]pyrrole-2,5-dione
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3C(=O)C=CC3=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 313.16
Percent Purity 95%
Chemical Name or Material 4-(Maleimidomethyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'