missing translation for 'onlineSavingsMsg'
Learn More

4-Ketopimelic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3778914
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
502-50-1
C7H10O5
174.15
MFCD00002798
UDDSEESQRGPVIL-UHFFFAOYSA-N
95084
4-oxoheptanedioic acid
OC(=O)CCC(=O)CCC(O)=O
This item is not returnable. View return policy
502-50-1
C7H10O5
174.15
MFCD00002798
UDDSEESQRGPVIL-UHFFFAOYSA-N
95084
4-oxoheptanedioic acid
OC(=O)CCC(=O)CCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 142.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH2COCH2CH2CO2H
Quantity 5 g
Beilstein 03, 804
Synonym 4-ketopimelic acid, 4-oxopimelic acid, 4-ketopimelic, .gamma.-ketopimelic acid, heptanedioic acid, 4-oxo, acmc-20amfs, pimelic acid, 4-keto, heptanedioic acid,4-oxo, 4-oxoheptanedioic acid
Formula Weight 174.15
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 4-Ketopimelic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ketopimelic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'