missing translation for 'onlineSavingsMsg'
Learn More

4-keto-4,5,6,7-tetrahydrothianaphthene 97%, ACROS Organics™

Brand:  Acros Organics 122890100

Code : 29

Additional Details : CAS Number : 13414-95-4 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10495603

  • £291.20 / 10g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Keto-4,5,6,7-tetrahydrothianaphthene
96.0
≥96.0%
C8H8OS
MFCD00005861
13414-95-4
100.0
97%
152.21

Specifications

4-Keto-4,5,6,7-tetrahydrothianaphthene
96.0
Authentic
≥96.0%
Glass bottle
152.21
White to yellow
10g
13414-95-4
100.0
Low Melting Solid or Liquid After Melting
97%
C8H8OS
MFCD00005861
34° to 40°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Keto-4,5,6,7-tetrahydrothianaphthene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

SDS