missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Isopropylsulfamoyl)benzeneboronic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(Isopropylsulfamoyl)benzeneboronic acid, is used as coupling catalyst used in the synthesis of a number of organic compounds. Also used as fine intermediate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-(Isopropylsulfamoyl)benzeneboronic acid, is used as coupling catalyst used in the synthesis of a number of organic compounds. Also used as fine intermediate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 136°C to 140°C |
Quantity | 250 mg |
Formula Weight | 243.08 |
Percent Purity | 98% |
Chemical Name or Material | 4-(Isopropylsulfamoyl)benzeneboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'