missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-isopropylphenyl isothiocyanate, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Density | 1.0300g/mL |
Assay Percent Range | 95%+ |
Packaging | Amber glass bottle |
Linear Formula | (CH3)2CHC6H4NCS |
Quantity | 1 g |
Formula Weight | 177.27 |
Percent Purity | 95% |
Chemical Name or Material | 4-Isopropylphenyl isothiocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'