missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Isopropylbenzonitrile, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Linear Formula | (CH3)2CHC6H4CN |
Quantity | 5 g |
Formula Weight | 145.2 |
Percent Purity | 95% |
Chemical Name or Material | 4-Isopropylbenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'