missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10686174
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
122-03-2
C10H12O
148.21
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
122-03-2
C10H12O
148.21
MFCD00006953
WTWBUQJHJGUZCY-UHFFFAOYSA-N
4-isopropylbenzaldehyde, cuminaldehyde, cuminic aldehyde, cumaldehyde, cuminal, p-cumic aldehyde, cumic aldehyde, cuminyl aldehyde, p-isopropylbenzaldehyde, benzaldehyde, 4-1-methylethyl
326
CHEBI:28671
4-propan-2-ylbenzaldehyde
CC(C)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9790g/mL
Boiling Point 234°C to 235°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5290 to 1.532
Quantity 2.5 kg
Merck Index 15, 2611
Specific Gravity 0.979
Solubility Information Solubility in water: insoluble
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Isopropylbenzaldehyde, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Isopropylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'