missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Isopropoxy-2-methylbenzeneboronic acid, 98%

Catalog Number p-7035415
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
871126-21-5
C10H15BO3
194.037
MFCD07363788
OEBXBUNQDNBWRP-UHFFFAOYSA-N
4-isopropoxy-2-methylphenylboronic acid, 4-isopropoxy-2-methylphenyl boronic acid, 4-isopropoxy-2-methylbenzeneboronic acid, boronic acid,b-2-methyl-4-1-methylethoxy phenyl, 2-methyl-4-propan-2-yloxy phenyl boronic acid, acmc-209qf7, 4-isopropoxy-2-methylphenyl boronicacid, 4-isopropoxy-2-methyl-phenyl boronic acid, 2-methyl-4-propan-2-yl oxy phenyl boronic acid
16218044
(2-methyl-4-propan-2-yloxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC(C)C)C)(O)O
This item is not returnable. View return policy
871126-21-5
C10H15BO3
194.037
MFCD07363788
OEBXBUNQDNBWRP-UHFFFAOYSA-N
4-isopropoxy-2-methylphenylboronic acid, 4-isopropoxy-2-methylphenyl boronic acid, 4-isopropoxy-2-methylbenzeneboronic acid, boronic acid,b-2-methyl-4-1-methylethoxy phenyl, 2-methyl-4-propan-2-yloxy phenyl boronic acid, acmc-209qf7, 4-isopropoxy-2-methylphenyl boronicacid, 4-isopropoxy-2-methyl-phenyl boronic acid, 2-methyl-4-propan-2-yl oxy phenyl boronic acid
16218044
(2-methyl-4-propan-2-yloxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC(C)C)C)(O)O
Melting Point 112°C to 116°C
Quantity 1 g
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 4-Isopropoxy-2-methylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'